missing translation for 'onlineSavingsMsg'
Learn More

ABL2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications

Brand:  Novus Biologicals NBP1-18875

Product Code. 18057595

  • £496.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ABL2 Polyclonal specifically detects ABL2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ABL2
Polyclonal
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Abelson murine leukemia viral (v-abl) oncogene homolog 2, Abelson murine leukemia viral oncogene homolog 2, Abelson-related gene protein, Abelson-relatedgene), arg tyrosine kinase, EC 2.7.10, EC 2.7.10.2, FLJ22224, FLJ31718, FLJ41441, tyrosine-protein kinase ABL2, Tyrosine-protein kinase ARG, v-abl Abelson murine leukemia viral oncogene homolog 2
Rabbit
128 kDa
100 μL
Protein Kinase
27
Human, Mouse
IgG
Western Blot, Immunoprecipitation
1.0 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
P42684
ABL2
The immunogen recognized by this antibody maps to a region between residue 750 and 800 of human Abelson-related gene protein using the numbering given in entry NP_009298.1 (GeneID 27).
Affinity Purified
RUO
Primary
Human. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Bovine, Dog, Horse, Guinea pig_10141, Pig, Panda, Rhesus Monkey, Gorilla, White-tufted-ear marmoset, Crab-eating macaque, African elephant, Chinese hamster, Naked mole rat and Northern white-cheeked gibbon.
Store at 4C. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only