missing translation for 'onlineSavingsMsg'
Learn More

ABL2 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18416944 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416944 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416944 Supplier Novus Biologicals Supplier No. NBP118875IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ABL2 Polyclonal antibody specifically detects ABL2 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABL2
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Abelson murine leukemia viral (v-abl) oncogene homolog 2, Abelson murine leukemia viral oncogene homolog 2, Abelson-related gene protein, Abelson-relatedgene), arg tyrosine kinase, EC 2.7.10, EC 2.7.10.2, FLJ22224, FLJ31718, FLJ41441, tyrosine-protein kinase ABL2, Tyrosine-protein kinase ARG, v-abl Abelson murine leukemia viral oncogene homolog 2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 750 and 800 of human Abelson-related gene protein using the numbering given in entry NP_009298.1 (GeneID 27).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 27
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.