missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ABL2 Monoclonal antibody specifically detects ABL2 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Proximity Ligation Assay, KnockDown
Specifications
Specifications
| Antigen | ABL2 |
| Applications | Western Blot, ELISA, Immunoprecipitation, Proximity Ligation Assay, KnockDown |
| Classification | Monoclonal |
| Clone | 6D5 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA, Immunoprecipitation, Proximity Ligation Assay, Knockdown Validated |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | AAH65912 |
| Gene Alias | Abelson murine leukemia viral (v-abl) oncogene homolog 2, Abelson murine leukemia viral oncogene homolog 2, Abelson-related gene protein, Abelson-relatedgene), arg tyrosine kinase, EC 2.7.10, EC 2.7.10.2, FLJ22224, FLJ31718, FLJ41441, tyrosine-protein kinase ABL2, Tyrosine-protein kinase ARG, v-abl Abelson murine leukemia viral oncogene homolog 2 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?