missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ABL1 [F317L], GST Tag Recombinant Protein

Product Code. 15940403
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
This item is not returnable. View return policy

Product Code. 15940403

Brand: Invitrogen™ A33475

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This ABL1 F317L recombinant human protein (27-end) was expressed in insect cells. ABL1 F317L recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Z'-LYTE™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Mutant The gene accession number for ABL1 [F317L] is NP_005148.2.

ABL1 cytoplasmic tyrosine kinase protooncogene encodes a cytoplasmic and nuclear kinase that has been implicated in processes of cell differentiation, cell division, cell adhesion, and stress response. Activity of ABL protein is negatively regulated by its SH3 domain and deletion of the SH3 domain turns ABL1 into an oncogene. Translocation and head-to-tail fusion of the BCR and ABL1 genes is present in many cases of chronic myelogeneous leukemia. The DNA-binding activity of the ubiquitously expressed ABL1 tyrosine kinase is regulated by CDK1-mediated phosphorylation, suggesting a cell cycle function for ABL1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P00519
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 10mM glutathione, 150mM NaCl, 0.25mM DTT, 0.1mM PMSF, 0.1mM EDTA, 25% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 25
Molecular Weight (g/mol) 146.3 kDa
Name Human ABL1 [F317L], GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias ABL1
Common Name ABL1 (F317L Mutant)
Gene Symbol ABL1
KinaseFamily ABL Kinase Family
Protein Length 27-end
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 27-end
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Active, Mutant
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.