missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX Abl (M351T) Protein, Active

Product Code. 17300674
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17300674

Brand: Eurofins DiscoverX 14757

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, N-terminal 6His-tagged recombinant human Abl, residues 27-end containing the M351T mutation, for use in Kinase Assays.

The abl oncogene was initially identified as the viral transforming gene (v-abl) of Abelson murine leukemia virus (A-MuLV), from a chemically thymectomized mouse inoculated with Moloney murine leukemia virus (M-MuLV). The v-abl genome consists of M-MuLV gag sequences fused to mouse c-abl, resulting in the production of phosphoroteins, p160 and p120 gag/c-abl with protein-tyrosine kinase activity, as the major translational products. The abl oncogene is implicated in serval human leukemias including chronic myelocytic (CML), adult acute lymphoblastic (ALL) and pediatric ALL. In these leukemias the c-abl proto-oncogene undergoes a chromosomal translocation producing the Philadelphia (Ph¢) chromosome resulting in the fusion of c-abl sequences to the bcr gene of chromosome 22, and the molecular consequences of this translocation is the generation of a chimeric bcr/c-abl mRNA encoding activated abl protein-tyrosine kinase.

GenBank U07563

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1 mM EGTA, 0.1% 2-mercaptoethanol, 0.03% Brij-35. Frozen solution.
Molecular Weight (g/mol) 121.4 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Truncated
Purity or Quality Grade ≥40%
Protein Abl (M351T), active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.