Learn More
Eurofins DiscoverX Abl (H396P) Protein, Active
Description
The abl oncogene was initially identified as the viral transforming gene (v-abl) of Abelson murine leukemia virus (A-MuLV), from a chemically thymectomized mouse inoculated with Moloney murine leukemia virus (M-MuLV). The v-abl genome consists of M-MuLV gag sequences fused to mouse c-abl, resulting in the production of phosphoroteins, p160 and p120 gag/c-abl with protein-tyrosine kinase activity, as the major translational products. The abl oncogene is implicated in serval human leukemias including chronic myelocytic (CML), adult acute lymphoblastic (ALL) and pediatric ALL. In these leukemias the c-abl proto-oncogene undergoes a chromosomal translocation producing the Philadelphia (Ph¢) chromosome resulting in the fusion of c-abl sequences to the bcr gene of chromosome 22, and the molecular consequences of this translocation is the generation of a chimeric bcr/c-abl mRNA encoding activated abl protein-tyrosine kinase.
GenBank U07563
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1 mM EGTA, 0.1% 2-mercaptoethanol, 0.03% Brij-35. Frozen solution. |
| Molecular Weight (g/mol) | 121.4 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥36% |
| Protein | Abl (H396P), active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.