missing translation for 'onlineSavingsMsg'
Learn More

Abiraterone acetate, MedChemExpress

Product Code. 16448198
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
2 g
5 g
Unit Size:
100mg
10mg
1g
200mg
2g
500mg
50mg
5g
This item is not returnable. View return policy

Product Code. 16448198

Brand: Medchem Express HY75054

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Abiraterone acetate (CB7630) is an oral, potent, selective, and irreversible inhibitor of CYP17A1 with antiandrogen activity. Abiraterone acetate is a prodrug form of Abiraterone (CB7598).

Abiraterone acetate (CB7630) is an oral, potent, selective, and irreversible inhibitor of CYP17A1 with antiandrogen activity. Abiraterone acetate is a prodrug form of Abiraterone (CB7598).

Specifications

Chemical Name or Material Abiraterone acetate
Physical Form Solid
CAS 154229-18-2
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C26H33NO2
For Use With (Application) Cancer-programmed cell death
Synonym CB7630
Solubility Information DMSO : 10 mg/mL (25.54 mM; Need ultrasonic)
SMILES O=C(O[C@@H]1CC2=CC[C@]3([H])[C@]([H])(CC[C@](C)(C(C4=CN=CC=C4)=CC5)[C@]35[H])[C@@]2(C)CC1)C
Molecular Weight (g/mol) 391.55
Formula Weight 391.55
Percent Purity 95.8%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.