missing translation for 'onlineSavingsMsg'
Learn More

Abietic Acid, >95%, TRC

Product Code. 30314227
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314227 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314227 Supplier Toronto Research Chemicals Supplier No. TRCA108203100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Abietic Acid (Sylvic Acid)
CAS 514-10-3
Purity Grade Notes HPLC
Molecular Formula C20 H30 O2
Quantity 100 mg
Synonym 1-Phenanthrenecarboxylic acid, 1,2,3,4,4a,4b,5,6,10,10a-decahydro-1,4a-dimethyl-7-(1-methylethyl)-, [1R-(1α,4aβ,4bα,10aα)]-, Abietic acid, (-)- (7CI), Podocarpa-7,13-dien-15-oic acid, 13-isopropyl- (8CI), (1R,4aR,4bR,10aR)-1,2,3,4,4a,4b,5,6,10,10a-Decahydro-1,4a-dimethyl-7-(1-methylethyl)-1-phenanthrenecarboxylic acid, (-)-Abietic acid, 7,13-Abietadien-18-oic acid, Abieta-7,13-diene-18-oic acid, Abietic acid, NSC 25149, Odomit B 10, Sylvic acid, ZAO, ZAO (natural product), l-Abietic acid
InChI Formula InChI=1S/C20H30O2/c1-13(2)14-6-8-16-15(12-14)7-9-17-19(16,3)10-5-11-20(17,4)18(21)22/h7,12-13,16-17H,5-6,8-11H2,1-4H3,(H,21,22)/t16-,17+,19+,20+/m0/s1
SMILES CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(=O)O)[C@H]2CC1
IUPAC Name (1R,4aR,4bR,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid
Molecular Weight (g/mol) 302.45
Formula Weight 302.2246
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.