missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abietic acid, 90+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
Abietic acid is used to make ester gums in lacquers, varnishes and soaps. It is also used in metal resonates and plastics. Further, it is utilized in musical instruments to make them less slippery. It acts as a precursor to natural product synthesis. In addition to this, it is used to depackage integrated circuits from their epoxy coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 148°C to 152°C |
Color | Yellow |
Beilstein | 2221451 |
Merck Index | 14,7 |
Solubility Information | Soluble in acetone,petroleum ether,diethyl ether and ethanol. Insoluble in water. |
Formula Weight | 302.45 |
Percent Purity | ≥90% |
Quantity | 250 mg |
Odor | Characteristic |
Physical Form | Crystalline |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'