missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ABI3 Recombinant Rabbit Monoclonal Antibody (3V2G1)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558083

 View more versions of this product

Product Code. 30283053

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: VPDGRLSAAS SAFSLASAGS AEGVGGAPTP KGQAAPPAPP LPSSLDPPPP PAAVEVFQRP PTLEELSPPP PDEELPLPLD LPPPPPLDGD ELGLPPPPPG FGPDEPSWVP ASYLEKVVTL YPYTSQKDNE LSFSEGTVIC VTRRYSDGWC EGVSSEGTGF FPGNYVEPSC.

Inhibits ectopic tumor cell metastasis of SRD cells. In vitro, reduces cell motility.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ABI3
Recombinant Monoclonal
1.08 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Q9P2A4
ABI3
Recombinant fusion protein containing a sequence corresponding to amino acids 197-366 of human ABI3 (NP_057512.2).
20 μL
Primary
Human
Antibody
IgG
ELISA, Western Blot
3V2G1
Unconjugated
ABI3
2210414K06Rik; ABI family member 3; ABI family, member 3; ABI gene family member 3; ABI gene family, member 3; Abi3; AI987680; NESH; NESH protein; new molecule including SH3; SSH3BP3
Rabbit
Protein A
RUO
51225
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.