missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABI2 Recombinant Rabbit Monoclonal Antibody (JE53-89)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA536165
This item is not returnable.
View return policy
Description
ABI2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
ABI2 may act in regulation of cell growth and transformation by interacting with nonreceptor tyrosine kinases ABL1 and/or ABL2. ABI2 is a part of the WAVE complex that regulates lamellipodia formation. The WAVE complex regulates actin filament reorganization via its interaction with the Arp2/3 complex. It regulates ABL1/c-Abl-mediated phosphorylation of MENA.
Specifications
| ABI2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P62484, Q9NYB9 | |
| Abi2 | |
| Recombinant protein within Human ABI2 aa 312-411. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| JE53-89 | |
| Unconjugated | |
| Abi2 | |
| 8430425M24Rik; Abelson interactor 2; Abi2; abi-2; ABI2B; abl binding protein 3; abl interactor 2; Abl-binding protein 3; AblBP3; abl-interacting protein 1 (SH3-containing protein); abl-interactor 2; abl-interactor protein 2b; AI839867; AIP-1; arg protein tyrosine kinase-binding protein; arg-binding protein 1; ArgBP1; ARGBPIA; argBPIB; C130078H13; SSH3BP2; thyroid hormone responsive protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10152, 329165 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction