missing translation for 'onlineSavingsMsg'
Learn More

ABI2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen ABI2
Concentration 0.41 mg/mL
Applications Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16843784
View Documents
Proteintech
14890-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16833784
View Documents
Proteintech
14890-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Interaction of Fas ligand (TNFSF6) with the FAS antigen (TNFRSF6) mediates programmed cell death, also called apoptosis, in a number of organ systems. The protein encoded by this gene binds to FAS antigen and can initiate apoptosis or enhance apoptosis initiated through FAS antigen. Initiation of apoptosis by the protein encoded by this gene requires a ubiquitin-like domain but not the FAS-binding domain.
Specifications

Specifications

ABI2
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Rat, Mouse
P62484, Q9NYB9
10152, 286928, 329165
ABI2 Fusion Protein Ag6679
Primary
-20°C
ABI2
0.41 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Abelson interactor 2, ABI 2, ABI2, ABI2B, Abl binding protein 3, abl interactor 2, AblBP3, AIP 1, Arg binding protein 1, ArgBP1, argBPIA, argBPIB, SSH3BP2
Abi2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.