missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABHDB Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562022
This item is not returnable.
View return policy
Description
Immunogen sequence: RFVWRVNLDA LTQHLDKILA FPQRQESYLG PTLFLLGGNS QFVHPSHHPE IMRLFPRAQM QTVPNAGHWI HADRPQDFIA AIRGFLV Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 70%.
ABHDB encodes a protein containing an alpha/beta hydrolase fold domain. This protein is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23.
Specifications
| ABHDB | |
| Polyclonal | |
| Unconjugated | |
| ABHD11 | |
| ABHD11; abhydrolase domain containing 11; abhydrolase domain-containing protein 11; Alpha/beta hydrolase domain-containing protein 11; PP1226; Protein ABHD11; WBSCR21; Williams Beuren syndrome chromosome region 21; Williams-Beuren syndrome chromosomal region 21 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83451 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NFV4 | |
| ABHD11 | |
| Recombinant protein corresponding to Human ABHDB. Recombinant protein control fragment (Product #RP-92658). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction