Learn More
ABHD6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20494-1-AP-150UL
 View more versions of this product
Product Code. 16880035
  • £349.00 / 150µL
Description
ABHD6 has 2-arachidonoylglycerol hydrolase activity. ABHD6 may be a regulator of endocannabinoid signaling pathways.
Specifications
Specifications
ABHD6
Polyclonal
Unconjugated
ABHD6
ABHD6, Monoacylglycerol lipase ABHD6
Rabbit
Antigen Affinity Chromatography
RUO
57406, 66082
-20°C
Liquid
Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8R2Y0, Q9BV23
Abhd6
ABHD6 Fusion Protein Ag13968
150 μL
Primary
Human, Mouse