missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABHD5 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67779-1-IG-20UL
This item is not returnable.
View return policy
Description
The ABHD5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ABHD5. This antibody recognizes human antigen. The ABHD5 antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| ABHD5 | |
| Monoclonal | |
| 1000 μg/mL | |
| CDS; CGI58; IECN2; MGC8731; NCIE2 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 39 kDa | |
| Liquid |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| 2B12C2 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 51099 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction