missing translation for 'onlineSavingsMsg'
Learn More

Abhd5 Antibody, Novus Biologicals™

Product Code. 18426720 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426720 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426720 Supplier Novus Biologicals Supplier No. NBP184507

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Abhd5 Polyclonal antibody specifically detects Abhd5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Abhd5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias abhydrolase domain containing 5, Abhydrolase domain-containing protein 5,1-acylglycerol-3-phosphate O-acyltransferase ABHD5, CDS, CGI58, EC 2.3.1.51, IECN2, Lipid droplet-binding protein CGI-58, MGC8731, NCIE2CGI-58
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: MTIPYGWAKRPMLQRIGKMHPDIPVSVIFGARSCIDGNSGTSIQSLRPHSYVKTIAILGAGHYVYADQPEEFNQKVKEI
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism, Lipid Droplets, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51099
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.