missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ABHD4 Polyclonal antibody specifically detects ABHD4 in Human samples. It is validated for ELISA
Specifications
Specifications
| Formulation | 50mM Sodium Borate |
| Gene Alias | ABH4, abhydrolase domain containing 4, abhydrolase domain-containing protein 4, Alpha/beta-hydrolase 4, EC 3.1.1.-, FLJ12816, Lyso-N-acylphosphatidylethanolamine lipase |
| Primary or Secondary | Primary |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?