missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABHD3 Monoclonal Antibody (A5D4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA544987
This item is not returnable.
View return policy
Description
ABHD3 Monoclonal Antibody for Western Blot
The α/β hydrolase superfamily comprise diverse members that are involved in important biochemical processes and related to various diseases. They have unrelated sequences, various substrates, and different kinds of catalytic activities, yet they share the same canonical α/β hydrolase fold, which consists of an eight stranded parallel α/β structure. They are also characterized by a catalytic triad composed of a histidine, an acid and a nucleophile. Members of this superfamily are often drug targets for treating diseases, such as diabetes, Alzheimer's disease, obesity and blood clotting disorders. The α/β hydrolase domain containing (ABHD) gene subfamily is comprised of 15 mostlyuncharacterized members, most of which utilize a serine nucleophile to form the G-X-S-X-G nucleophile elbow. ABHD1 plays a role in metabolizing smoking xenobiotics. ABHD2 participates in the development of atherosclerosis. ABHD3 is a 409 amino acid single-pass type II membrane protein. ABHD4 is involved in an alternative synthesis pathway of NAE (N-acyl ethanolamine). Mutations in ABHD5 contribute to Chanarin-Dorfman syndrome. ABDH6 may play a role in nervous system metabolism and signaling.
Specifications
| ABHD3 | |
| Monoclonal | |
| 2 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Plant | |
| Antibody | |
| IgG1 |
| Western Blot | |
| A5D4 | |
| Unconjugated | |
| AA675331; ABHD3; abhydrolase domain containing 3; abhydrolase domain-containing protein 3; alpha/beta hydrolase domain containing protein 3; LABH3; Lung alpha/beta hydrolase 3; mmLABH3; Phospholipase ABHD3 | |
| Recombinant full length protein within Camellia. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction