missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABHD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14039-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein containing an alpha/beta hydrolase fold, which is a catalytic domain found in a very wide range of enzymes. The function of this protein has not been determined. Alternative splicing of this gene results in two transcript variants encoding the same protein.Specifications
| ABHD2 | |
| Polyclonal | |
| Unconjugated | |
| ABHD2 | |
| ABHD2, HS1 2, LABH2, Lung alpha/beta hydrolase 2, PHPS1 2, Protein PHPS1 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11057, 293050, 54608 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P08910, Q9QXM0 | |
| Abhd2 | |
| ABHD2 Fusion Protein Ag5108 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction