missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABHD17A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140126
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LGTLRASSGA PGRWKLHLTE RADFQYSQ Highest antigen sequence indentity to the following orthologs: Rat - 71%, Mouse - 71%.
Hydrolyzes fatty acids from S-acylated cysteine residues in proteins (PubMed:26701913). Has depalmitoylating activity towards NRAS (PubMed:26701913). Has depalmitoylating activity towards DLG4/PSD95 (PubMed:26701913). May have depalmitoylating activity towards MAP6. [UniProt].
Specifications
| ABHD17A | |
| Polyclonal | |
| Unconjugated | |
| ABHD17A | |
| 1700013O15Rik; Abhd17a; abhydrolase domain containing 17A; abhydrolase domain containing 17A, depalmitoylase; abhydrolase domain-containing protein 17A; abhydrolase domain-containing protein FAM108A; abhydrolase domain-containing protein FAM108A1; alpha/beta hydrolase domain-containing protein 17A; BC005632; C19orf27; D10Bwg1364e; FAM108A; FAM108A1; family with sequence similarity 108, member A; family with sequence similarity 108, member A1; mFLJ00358 protein; MGC138013 protein; protein ABHD17A; RGD1359682; upf0227 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 81926 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96GS6 | |
| ABHD17A | |
| Recombinant protein corresponding to Human ABHD17A. Recombinant protein control fragment (Product #RP-109180). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction