missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABHD15 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22526-1-AP-20UL
This item is not returnable.
View return policy
Description
ABHD15 gene ontology annotations related to this gene include cellular lipid metabolic process.Specifications
| ABHD15 | |
| Polyclonal | |
| Unconjugated | |
| ABHD15 | |
| ABHD15 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116236, 67477 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5F2F2, Q6UXT9 | |
| Abhd15 | |
| ABHD15 Fusion Protein Ag18334 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction