missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABHD14B Rabbit anti-Human, Clone: 2, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | ABHD14B |
|---|---|
| Clone | 2 |
| Dilution | ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunofluorescence |
| Applications | ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18216686
|
Novus Biologicals
NBP2-90259-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18204098
|
Novus Biologicals
NBP2-90259-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ABHD14B Monoclonal specifically detects ABHD14B in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| ABHD14B | |
| ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 84836 | |
| IgG | |
| Protein A purified |
| 2 | |
| ELISA, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| abhydrolase domain containing 14B, abhydrolase domain-containing protein 14B, CCG1-interacting factor B, CIBcell cycle gene 1-interacting factor B, EC 3.-, EC 3.1.11.6, EC 3.1.21.4, MGC15429 | |
| Recombinant Human ABHD14B Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title