missing translation for 'onlineSavingsMsg'
Learn More

ABHD14B Rabbit anti-Human, Clone: 11, Novus Biologicals™

Rabbit Monoclonal Antibody

£286.00 - £429.00

Specifications

Antigen ABHD14B
Clone 11
Dilution Western Blot 1:500-1:1000, Immunoprecipitation 0.2-1 ul/mg of lysate
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18261356
View Documents
Novus Biologicals
NBP2-90260-50UL
50 μg
£286.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18227167
View Documents
Novus Biologicals
NBP2-90260-100UL
100 μg
£429.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ABHD14B Monoclonal specifically detects ABHD14B in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

ABHD14B
Western Blot 1:500-1:1000, Immunoprecipitation 0.2-1 ul/mg of lysate
Monoclonal
Purified
RUO
0.2 um filtered solution in PBS
84836
IgG
Protein A purified
11
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Human
abhydrolase domain containing 14B, abhydrolase domain-containing protein 14B, CCG1-interacting factor B, CIBcell cycle gene 1-interacting factor B, EC 3.-, EC 3.1.11.6, EC 3.1.21.4, MGC15429
Recombinant Human ABHD14B protein
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.