missing translation for 'onlineSavingsMsg'
Learn More

ABHD14B Antibody, DyLight 488, Novus Biologicals™

Product Code. 18322223 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322223 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18322223 Supplier Novus Biologicals Supplier No. NBP300217G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ABHD14B Polyclonal specifically detects ABHD14B in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABHD14B
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias abhydrolase domain containing 14B, abhydrolase domain-containing protein 14B, CCG1-interacting factor B, CIBcell cycle gene 1-interacting factor B, EC 3.-, EC 3.1.11.6, EC 3.1.21.4, MGC15429
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant ABHD14B (Q96IU4-1; Met1-Gln210).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 84836
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.