missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ABHD12B Polyclonal specifically detects ABHD12B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | ABHD12B |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | Q7Z5M8 |
| Gene Alias | abhydrolase domain containing 12B, abhydrolase domain-containing protein 12B, BEM46L3, c14_5314, C14orf29, chromosome 14 open reading frame 29, EC 3.-, MGC129926, MGC129927 |
| Gene Symbols | ABHD12B |
| Host Species | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?