missing translation for 'onlineSavingsMsg'
Learn More

ABHD12 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™

Product Code. 18366564 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18366564 20 μg 20µL
18399883 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18366564 Supplier Novus Biologicals Supplier No. NBP31554920UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ABHD12 Polyclonal antibody specifically detects ABHD12 in Human, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABHD12
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias ABHD12A, abhydrolase domain containing 12, Abhydrolase domain-containing protein 12, BEM46L2, C20orf22, chromosome 20 open reading frame 22, dJ965G21.2, DKFZP434P106, DKFZp434P106,2-arachidonoylglycerol hydrolase, EC 3.1.1.23, monoacylglycerol lipase ABHD12, PHARC
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 110-250 of human ABHD12 (NP_056415.1).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26090
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.