missing translation for 'onlineSavingsMsg'
Learn More

ABHD12 Antibody, Novus Biologicals™

Product Code. 18364840 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364840 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364840 Supplier Novus Biologicals Supplier No. NBP286941

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ABHD12 Polyclonal specifically detects ABHD12 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABHD12
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias ABHD12A, abhydrolase domain containing 12, Abhydrolase domain-containing protein 12, BEM46L2, C20orf22, chromosome 20 open reading frame 22, dJ965G21.2, DKFZP434P106, DKFZp434P106,2-arachidonoylglycerol hydrolase, EC 3.1.1.23, monoacylglycerol lipase ABHD12, PHARC
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C-terminal region of ABHD12
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26090
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.