missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABH2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56921
This item is not returnable.
View return policy
Description
ABH2 Polyclonal specifically detects ABH2 in Human samples. It is validated for Western Blot.
Specifications
| ABH2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| 2OG-Fe(II) oxy DC1, ABH2FLJ99103, alkB, alkylation repair homolog 2 (E. coli), Alkylated DNA repair protein alkB homolog 2, alpha-ketoglutarate-dependent dioxygenase alkB homolog 2, EC 1.14.11.-, MGC90512, Oxy DC1 | |
| Rabbit | |
| 29 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Pig: 100%; Rat: 100%; Canine: 92%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q6NS38 | |
| ALKBH2 | |
| Synthetic peptides corresponding to ALKBH2(alkB, alkylation repair homolog 2 (E. coli)) The peptide sequence was selected from the middle region of ABH2 (NP_001001655). Peptide sequence VPRKQATYGDAGLTYTFSGLTLSPKPWIPVLERIRDHVSGVTGQTFNFVL. | |
| Affinity purified | |
| RUO | |
| 121642 | |
| Centrifuge prior to reconstitution. Reconstitute with 50μL distilled water to a final antibody concentration of 1mg/mL. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction