missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABH1 Recombinant Rabbit Monoclonal Antibody (23GB1385), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | ABH1 |
|---|---|
| Clone | 23GB1385 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088613
![]() |
Invitrogen™
MA554170 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088903
|
Invitrogen™
MA554171 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ABH1 Recombinant Monoclonal antibody specifically detects ABH1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| ABH1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q13686 | |
| 8846 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1385 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2700073G19Rik; Abh; ABH1; alkB; alkB homolog 1, histone H2A dioxygenase; alkB, alkylation repair homolog 1; alkB, alkylation repair homolog 1 (E. coli); ALKBH; Alkbh1; Alkylated DNA repair protein alkB homolog 1; alkylation repair, alkB homolog; alpha-ketoglutarate-dependent dioxygenase ABH1; DNA 6mA demethylase; DNA demethylase ALKBH1; DNA lyase ABH1; DNA N6-methyl adenine demethylase; DNA N6-methyl adenine demethylase ALKBH1; DNA oxidative demethylase ALKBH1; hABH; mRNA N(3)-methylcytidine demethylase; Nucleic acid dioxygenase ALKBH1; tRNA N1-methyl adenine demethylase | |
| A synthesized peptide derived from ALKBH1 (1-85AA). | |
| Primary | |
| -20°C | |
| ALKBH1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title