Learn More
Thermo Scientific™ Abgene™ 2D Barcoded Twist-Lock Storage Tubes
Product Code. 12572627
Click to view available options
Product Type:
Screw Top Tube
Unit Size:
Pack of 10
Product Code. 12572627
Brand: Thermo Scientific™ AB1248
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Get 2D Barcoded Twist-Lock Storage Tubes for the perfect solution for sample storage in most laboratory conditions down to -80°C.
Encapsulated onto the base of every Thermo Scientific Abgene Barcoded Twist-Lock Storage Tube is a unique 2D barcode to securely identify and track samples, even in the harshest of laboratory conditions. Instantly decode each tube’s 2D barcode with the Thermo Scientific line of 2D barcode readers; scan the tube into any application or database, or integrate via various data export options-the choice is yours. Once sample data is stored, just scan the tube using the database ‘find’ feature to instantly access sample information and identify the contents of each storage tube.
Secure Tracking
  • 2D barcode‚ is visible‚ even in the harshest of laboratory conditions
  • Instantly decode each tube's 2D barcode with the Thermo Scientific line of 2D barcode readers; scan the tube into any application or database, or integrate via various data export options
  • Once sample data is stored, scan the tube using the database Find feature to instantly access sample information and identify the contents of each storage tube
Superior Design
  • Save precious freezer space with an innovative, stackable, 6 x 8 microplate-footprint rack of 48 tubes with a familiar cryopreservation format
  • Alphanumeric grid-referencing on rack and lid clearly display the storage tube location
  • Externally threaded tubes prevent contamination
  • Open rack bottom allows all barcodes to be read at once, saving time and allowing more efficient freezing and thawing
  • Unlike other plastic or cardboard storage boxes, the stackable, secure rack also ensures your freezer stays organized for faster sample retrieval
  • V-shaped internal well design reduces dead volume and improves sample recovery
Stringent Quality Control
  • Each tube is scanned to ensure readability
  • Each code is checked against a full database of all previously assigned 2Dcodes to ensure zero duplicates across the entire line of Abgene 2D barcoded storage tubes
  • Each lot of storage tubes is leak tested to ensure the integrity and security of samples
Recommended for:
Individual sample retrieval; Instant sample identification; Secure identification of archived samples; Secure shipping of samples; Storage of genomic samples, DMSO dissolved compounds; Tracking of samples during manufacturing
TRUSTED_SUSTAINABILITY
Specifications
Product Type Screw Top Tube
Description 2D Cypher Twist-Lock tubes
Capacity (Metric) 1.0 mL
Barcode 2D Barcode
Packaging Quantity 5 Cs./Pk.
Capacity (English) 0.0338 fl oz.