missing translation for 'onlineSavingsMsg'
Learn More

Abemaciclib methanesulfonate, MedChemExpress

Product Code. p-200098364
Encompass_Preferred
Click to view available options
Quantity:
5 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16481216

Brand: Medchem Express HY16297

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Abemaciclib methanesulfonate (LY2835219 methanesulfonate) is a selective CDK4/6 inhibitor with IC50s of 2 nM and 10 nM for CDK4 and CDK6, respectively.

Abemaciclib methanesulfonate (LY2835219 methanesulfonate) is a selective CDK4/6 inhibitor with IC50s of 2 nM and 10 nM for CDK4 and CDK6, respectively.

Specifications

Chemical Name or Material Abemaciclib methanesulfonate
CAS 1231930-82-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C28H36F2N8O3S
Quantity 200 mg
Synonym LY2835219 methanesulfonate
Solubility Information H2O : 125 mg/mL (207.40 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (16.59 mM; ultrasonic and warming and heat to 80°C)
SMILES CC(N1C2=CC(C3=NC(NC4=NC=C(CN5CCN(CC)CC5)C=C4)=NC=C3F)=CC(F)=C2N=C1C)C.CS(=O)(O)=O
Molecular Weight (g/mol) 602.7
Formula Weight 602.7
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.