missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCG2/CD338 Rabbit anti-Human, Clone: 2K8X1, Novus Biologicals™
Rabbit Monoclonal Antibody
£167.00 - £406.00
Specifications
| Antigen | ABCG2/CD338 |
|---|---|
| Clone | 2K8X1 |
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18064135
|
Novus Biologicals
NBP3-15559-20UL |
20 μg |
£167.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18099495
|
Bio-Techne
NBP3-15559-100UL |
100 μg |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ABCG2/CD338 Monoclonal antibody specifically detects ABCG2/CD338 in Human samples. It is validated for Western BlotSpecifications
| ABCG2/CD338 | |
| Western Blot 1:500 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ABC transporter, ABC15, ABCPMGC102821, ATP-binding cassette sub-family G member 2, ATP-binding cassette transporter G2, ATP-binding cassette, sub-family G (WHITE), member 2, BCRP1, BCRPMRX, BMDP, Breast cancer resistance protein, CD338, CD338 antigen, CDw338, EST157481, mitoxantrone resistance protein, Mitoxantrone resistance-associated protein, MXRMXR1, placenta specific MDR protein, Placenta-specific ATP-binding cassette transporter | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human ABCG2/CD338 (Q9UNQ0). SGLSGDVLINGAPRPANFKCNSGYVVQDDVVMGTLTVRENLQFSAALRLATTMTNHEKNERINRVIQELGLDKVADSKVGTQFIRGVSGGERKRTSIGMEL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 2K8X1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| ABC Transporters, Cancer, Cellular Markers, Hematopoietic Stem Cell Markers, Neuronal Cell Markers, Stem Cell Markers, Stem Cells | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 9429 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title