missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCG2/CD338 Antibody (MM0047-2J39), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-93511AF647
This item is not returnable.
View return policy
Description
ABCG2/CD338 Monoclonal specifically detects ABCG2/CD338 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ABCG2/CD338 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ABCG2 | |
| Human recombinant ABCG2/CD338 | |
| 0.1 mL | |
| ABC Transporters, Cancer, Cellular Markers, Hematopoietic Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers, Stem Cells | |
| 9429 | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0047-2J39 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ABC transporter, ABC15, ABCPMGC102821, ATP-binding cassette sub-family G member 2, ATP-binding cassette transporter G2, ATP-binding cassette, sub-family G (WHITE), member 2, BCRP1, BCRPMRX, BMDP, Breast cancer resistance protein, CD338, CD338 antigen, CDw338, EST157481, mitoxantrone resistance protein, Mitoxantrone resistance-associated protein, MXRMXR1, placenta specific MDR protein, Placenta-specific ATP-binding cassette transporter | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This was selected for its ability to detect human ABCG2 by flow cytometry and immunocytochemistry. | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction