missing translation for 'onlineSavingsMsg'
Learn More

ABCG2/CD338 Antibody (3G8), DyLight 680, Novus Biologicals™

Product Code. 18408935 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408935 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408935 Supplier Novus Biologicals Supplier No. NBP222124FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ABCG2/CD338 Monoclonal antibody specifically detects ABCG2/CD338 in Human, Mouse, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCG2/CD338
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3G8
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias ABC transporter, ABC15, ABCPMGC102821, ATP-binding cassette sub-family G member 2, ATP-binding cassette transporter G2, ATP-binding cassette, sub-family G (WHITE), member 2, BCRP1, BCRPMRX, BMDP, Breast cancer resistance protein, CD338, CD338 antigen, CDw338, EST157481, mitoxantrone resistance protein, Mitoxantrone resistance-associated protein, MXRMXR1, placenta specific MDR protein, Placenta-specific ATP-binding cassette transporter
Host Species Mouse
Immunogen Purified recombinant fragment of human ABCG2 expressed in E. coli. [Uniprot: Q9UNQ0]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Cancer, Cellular Markers, Hematopoietic Stem Cell Markers, Neuronal Cell Markers, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 9429
Target Species Human, Mouse, Primate
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.