missing translation for 'onlineSavingsMsg'
Learn More

ABCF2 Antibody, FITC, Novus Biologicals™

Product Code. 18336932 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18336932 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18336932 Supplier Novus Biologicals Supplier No. NBP298976F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ABCF2 Polyclonal specifically detects ABCF2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCF2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias ABC28, ABC-type transport protein, ATP-binding cassette sub-family F member 2, ATP-binding cassette, sub-family F (GCN20), member 2, DKFZp586K1823, EC 3.6.3, EC 3.6.3.17, EST133090, HUSSY18, HUSSY-18, Iron inhibited ABC transporter 2, Iron-inhibited ABC transporter 2, M-ABC1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human ABCF2.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Chromatin Research, DNA replication Transcription Translation and Splicing, Lipid and Metabolism, Membrane Trafficking and Chaperones, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10061
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.