missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13950-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a component of the spliceosome complex and is one of several retinitis pigmentosa-causing genes. When the gene product is added to the spliceosome complex, activation occurs.Specifications
| ABCF1 | |
| Polyclonal | |
| Unconjugated | |
| ABCF1 | |
| ABC27, ABC50, ABCF1, ATP binding cassette 50 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 224742, 23, 85493 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6MG08, Q6P542, Q8NE71 | |
| Abcf1 | |
| ABCF1 Fusion Protein Ag5061 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction