missing translation for 'onlineSavingsMsg'
Learn More

ABCF1 Antibody (1B4), Novus Biologicals™

Product Code. 18354058 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354058 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18354058 Supplier Novus Biologicals Supplier No. H00000023M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ABCF1 Monoclonal antibody specifically detects ABCF1 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCF1
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 1B4
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001081
Gene Alias ABC27, ABC50TNFalpha-inducible ATP-binding protein, ATP-binding cassette 50, ATP-binding cassette 50 (TNF-alpha stimulated), ATP-binding cassette sub-family F member 1, ATP-binding cassette, sub-family F (GCN20), member 1, EC 2.7.7.6, EC 3.6.3.17, EST123147, TNF-alpha-stimulated ABC protein
Host Species Mouse
Immunogen ABCF1 (NP_001081, 642 a.a. ~ 739 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GEMRKNHRLKIGFFNQQYAEQLRMEETPTEYLQRGFNLPYQDARKCLGRFGLESHAHTIQICKLSGGQKARVVFAELACREPDVLILDEPTNNLDIES
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 23
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.