missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ABCE1 Recombinant Rabbit Monoclonal Antibody (K01_1K86)

Product Code. 30828193
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828193 50 μL
30825393 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828193 Supplier Invitrogen™ Supplier No. MA564626
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

ABCE1 Recombinant Monoclonal Antibody for Western Blot

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the OABP subfamily. Alternatively referred to as the RNase L inhibitor, this protein functions to block the activity of ribonuclease L. Activation of ribonuclease L leads to inhibition of protein synthesis in the 2-5A/RNase L system, the central pathway for viral interferon action. Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCE1
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1K86
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene ABCE1
Gene Accession No. P61221, P61222
Gene Alias 2'-5'-oligoadenylate-binding protein; ABC 38; ABC38; ABCE 1; ABCE1; ATP binding cassette subfamily E member 1; ATP-binding cassette sub-family E member 1; ATP-binding cassette, subfamily E (OABP), member 1; ATP-binding cassette, sub-family E (OABP), member 1; ATP-binding cassette, subfamily E, member 1; C79080; HP68; HuHP68; OABP; OK/SW-cl.40; Ribonuclease 4 inhibitor; ribonuclease L (2', 5'-oligoisoadenylate synthetase-dependent) inhibitor; ribonuclease L (2',5'-oligoisoadenylate synthetase-dependent) inhibitor; RLI; RLI1; RNase 4 inhibitor; RNase L inhibitor; RNase L inhibitor 1; RNASEL1; Rnaseli; RNS41; RNS4I; RNS4l (Eye)
Gene Symbols ABCE1
Host Species Rabbit
Immunogen A synthetic peptide of human ABCE1.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24015, 361390, 6059
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.