missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCD1 Mouse anti-Human, Clone: 1H8E5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60153-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene is a member of the UBA domain family, whose members include proteins having connections to ubiquitin and the ubiquitination pathway. The ubiquitin associated domain is thought to be a non-covalent ubiquitin binding domain consisting of a compact three helix bundle. This particular protein originates from a gene locus in a refined region on chromosome 9 undergoing loss of heterozygosity in nasopharyngeal carcinoma (NPC). Taking into account its cytogenetic location, this UBA domain family member is being studies as a putative target for mutation in nasopharyngeal carcinomas. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| ABCD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P33897 | |
| ABCD1 | |
| ABCD1 Fusion Protein Ag12757 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| 1H8E5 | |
| Unconjugated | |
| ABCD1 | |
| ABC42, ABCD1, Adrenoleukodystrophy protein, ALD, ALDP, AMN | |
| Mouse | |
| Caprylic Acid Precipitation | |
| RUO | |
| 215 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction