missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCD1 Antibody (OTI2C12), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70150AF532
This item is not returnable.
View return policy
Description
ABCD1 Monoclonal specifically detects ABCD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated, Knockdown Validated.
Specifications
| ABCD1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ABCD1 | |
| Recombinant protein fragment corresponding to amino acids 508-745 of human ABCD1 (NP_000024) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunoassay, KnockDown | |
| OTI2C12 | |
| Western Blot, Immunohistochemistry, Knockout Validated, Knockdown Validated | |
| Adrenoleukodystrophy protein, ALDATP-binding cassette sub-family D member 1, ALDPadrenoleukodystrophy, AMNABC42, ATP-binding cassette, sub-family D (ALD), member 1 | |
| Mouse | |
| Affinity Purified | |
| ABC Transporters, Lipid and Metabolism | |
| 215.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction