missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcc9 Mouse anti-Human, Mouse, Rat, APC, Clone:S319A-14, Abnova™
Mouse monoclonal antibody raised against partial recombinant mouse Abcc9.
Brand: Abnova MAB17051.100ug
This item is not returnable.
View return policy
Specifications
| Abcc9 | |
| Monoclonal | |
| APC | |
| Immunocytochemistry Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
| Abcc9 | |
| Mouse | |
| 100 μg | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| S319A-14 | |
| ATP-binding cassette, sub-family C (CFTR/MRP), member 9 | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| AI414027, AI449286, SUR2A, SUR2B, Sur2 | |
| Recombinant protein corresponding to amino acids 1505-1546 at C-terminus of mouse Abcc9. | |
| 20928 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction