missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCC5 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29792-1-AP-20UL
This item is not returnable.
View return policy
Description
The ABCC5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ABCC5. This antibody recognizes human antigen. The ABCC5 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| ABCC5 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 10057 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 600 μg/mL | |
| ABC33; DKFZp686C1782; EST277145; MOAT-C; MOATC; MRP5; SMRP; pABC11 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 161-200 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction