missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ABCC5 Antibody (1B12), Novus Biologicals™

Product Code. 18348069 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348069 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18348069 Supplier Novus Biologicals™ Supplier No. H00010057M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ABCC5 Monoclonal antibody specifically detects ABCC5 in Human samples. It is validated for ELISA,Western Blot,Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCC5
Applications ELISA, Western Blot, Sandwich ELISA
Classification Monoclonal
Clone 1B12
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005679
Gene Alias ATP-binding cassette sub-family C member 5, ATP-binding cassette, sub-family C (CFTR/MRP), member 5, canalicular multispecific organic anion transporter C, EST277145, MOAT-CABC33, MRP5MOATC, multidrug resistance-associated protein 5, Multi-specific organic anion transporter C, pABC11, SMRPDKFZp686C1782
Host Species Mouse
Immunogen ABCC5 (NP_005679.2, 760 a.a. ∽ 846 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. CITERGTHEELMNLNGDYATIFNNLLLGETPPVEINSKKETSGSQKKSQDKGPKTGSVKKEKAVKPEEGQLVQLEEKGQGSVPWSVY
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline ABC Transporters, Amino Acids Drugs and other small molecules
Primary or Secondary Primary
Gene ID (Entrez) 10057
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.