missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCC3, Mouse anti-Human, Clone: M3II-9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB3384.500uL
This item is not returnable.
View return policy
Specifications
| ABCC3 | |
| Monoclonal | |
| Unconjugated | |
| In serum-free culture supernatant (0.7% BSA, 0.09% sodium azide) | |
| ABC31/EST90757/MLP2/MOAT-D/MRP3/cMOAT2 | |
| Mouse | |
| 500 μL | |
| Primary | |
| M3II-9 reacts with an internal epitope of human MRP-3, a 190-200 kD transmembrane protein that is closely related to the multidrug resistance protein MRP1. Does not cross-react with human MDR1, MRP1, MRP2 or MRP-5 gene products. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
| M3II-9 | |
| Immunocytochemistry (1:20-1:50) Immunohistochemistry (Frozen sections) (1:20) Western Blot (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| ATP-binding cassette, sub-family C (CFTR/MRP), member 3 | |
| ABCC3 | |
| Recombinant fusion protein corresponding to amino acids 830-949 of human ABCC3. | |
| RUO | |
| 8714 | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction