missing translation for 'onlineSavingsMsg'
Learn More

ABCB9 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18427718 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427718 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427718 Supplier Novus Biologicals Supplier No. NBP297176AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ABCB9 Polyclonal antibody specifically detects ABCB9 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCB9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias ABC transporter 9 protein, ATP-binding cassette sub-family B member 9, ATP-binding cassette transporter 9, ATP-binding cassette, sub-family B (MDR/TAP), member 9, EC 3.6.3, EC 3.6.3.44, EST122234, hABCB9, KIAA1520, TAPL, TAP-like protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ABCB9 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 23457
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.