missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABCB6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542133
This item is not returnable.
View return policy
Description
Peptide sequence: VTEWRTKFRR AMNTQENATR ARAVDSLLNF ETVKYYNAES YEVERYREAI Sequence homology: Cow: 93%; Dog: 100%; Goat: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies. This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance as well as antigen presentation. This half-transporter likely plays a role in mitochondrial function. Localized to 2q26, this gene is considered a candidate gene for lethal neonatal metabolic syndrome, a disorder of mitochondrial function.
Specifications
| ABCB6 | |
| Polyclonal | |
| Unconjugated | |
| ABCB6 | |
| 1200005B17Rik; ABC; ABC14; ABCB6; ATG9 autophagy related 9 homolog A; ATP binding cassette subfamily B member 6; ATP binding cassette subfamily B member 6 (Langereis blood group); ATP-binding cassette half-transporter; ATP-binding cassette sub-family B member 6, mitochondrial; ATP-binding cassette, subfamily B (MDR/TAP), member 6; ATP-binding cassette, sub-family B (MDR/TAP), member 6; ATP-binding cassette, sub-family B (MDR/TAP), member 6 (Langereis blood group); ATP-binding cassette, sub-family B, member 6; DUH3; EST45597; FLJ22414; LAN; LOW QUALITY PROTEIN: ATP-binding cassette sub-family B member 6, mitochondrial; MCOPCB7; Mitochondrial ABC transporter 3; mitochondrial ATP-binding cassette sub-family B member 6; mt-ABC transporter 3; MTABC3; P-glycoprotein-related protein; PRP; PSHK2; ubiquitously-expressed mammalian ABC half transporter; Umat | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10058 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NP58 | |
| ABCB6 | |
| Synthetic peptide directed towards the middle region of human ABCB6. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction