missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCB5 Polyclonal Antibody

ABCB5 Polyclonal Antibody
Brand: Invitrogen PA513114
This item is not returnable.
View return policy
Description
ABCB5 Polyclonal Antibody for Western Blot, Flow
Specifications
| ABCB5 | |
| Polyclonal | |
| Unconjugated | |
| ABCB5 | |
| 9230106F14Rik; ABCB5; ABCB5 P-gp; ABCB5alpha; ABCB5beta; ATP binding cassette subfamily B member 5; ATP-binding cassette protein; ATP-binding cassette sub-family B member 5; ATP-binding cassette, subfamily B (MDR/TAP), member 5; ATP-binding cassette, sub-family B (MDR/TAP), member 5; EST422562; P-glycoprotein ABCB5; RGD1566342 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 340273, 77706 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| B5X0E4, Q2M3G0 | |
| ABCB5 | |
| KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human ABCB5. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction