missing translation for 'onlineSavingsMsg'
Learn More

ABCB5 Antibody, HRP, Novus Biologicals™

Product Code. 18405920
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405920 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405920 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP177687H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ABCB5 Polyclonal antibody specifically detects ABCB5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCB5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias ABCB5 P-gp, ABCB5alpha, ABCB5beta, ATP-binding cassette protein, ATP-binding cassette sub-family B member 5, ATP-binding cassette, sub-family B (MDR/TAP), member 5, EC 3.6.3, EC 3.6.3.44, EST422562, P-glycoprotein ABCB5
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human ABCB5 protein (between residues 150-200) [UniProt Q2M3G0]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 340273
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.