missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCB5 Antibody (CL13618) [Unconjugated], Novus Biologicals™
Mouse Monoclonal Antibody
£321.00 - £459.00
Specifications
| Antigen | ABCB5 |
|---|---|
| Clone | CL13618 |
| Concentration | 1 mg/mL |
| Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30279777
|
Novus Biologicals
NBP3-43642-25ul |
25 μL |
£321.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30279563
|
Novus Biologicals
NBP3-43642-100ul |
100 μL |
£459.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ABCB5 Monoclonal antibody specifically detects ABCB5 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)Specifications
| ABCB5 | |
| 1 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| ABC Transporters, Lipid and Metabolism | |
| PBS, pH 7.2, 40% glycerol | |
| 340273 | |
| IgG1 | |
| Protein A purified |
| CL13618 | |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ABCB5 P-gp, ABCB5alpha, ABCB5beta, ATP-binding cassette protein, ATP-binding cassette sub-family B member 5, ATP-binding cassette, sub-family B (MDR/TAP), member 5, EC 3.6.3, EC 3.6.3.44, EST422562, P-glycoprotein ABCB5 | |
| This antibody was generated using a synthetic peptide of Q2M3G0, with the exact immunogen sequence remaining proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title