missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCB5 Antibody (Brigham and Women's patent anti-ABCB5) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28518JF635
This item is not returnable.
View return policy
Description
ABCB5 Monoclonal antibody specifically detects ABCB5 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| ABCB5 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| ABCB5 P-gp, ABCB5alpha, ABCB5beta, ATP-binding cassette protein, ATP-binding cassette sub-family B member 5, ATP-binding cassette, sub-family B (MDR/TAP), member 5, EC 3.6.3, EC 3.6.3.44, EST422562, P-glycoprotein ABCB5 | |
| ABCB5 | |
| 0.1 mL | |
| ABC Transporters, Lipid and Metabolism | |
| 340273 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Brigham and Women's patent anti-ABCB5 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction